siRNA Details
| Gene Symbol: MTMR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004685.3 | NP_004676.3 | 1, 2 | 342 | ||
| XM_011535306.1 | XP_011533608.1 | 1, 2 | 288 | |||
| XM_011535307.1 | XP_011533609.1 | 1, 2 | 288 | |||
| XM_017020845.1 | XP_016876334.1 | 1, 2 | 288 | |||
| GenBank mRNA | AF406619.1 | - | 1, 2 | 23 | ||
| AK314587.1 | - | 1, 2 | 207 | |||
| BC040012.1 | - | 1, 2 | 214 | 5495564 | ||

MTMR6
Genomic Map