siRNA Details
| Gene Symbol: MTMR7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004686.4 | NP_004677.3 | 2 | 316 | ||
| XM_017013956.1 | XP_016869445.1 | 2 | 138 | |||
| GenBank mRNA | AK294468.1 | - | 2 | 77 | ||
| AK316283.1 | - | 2 | 136 | |||
| BC130577.1 | - | 2 | 102 | 40146608 | ||
| BC130579.1 | - | 2 | 102 | 40146610 | ||
| CR749240.1 | - | 2 | 114 | |||
| DC325464.1 | - | 2 | 316 | |||

MTMR7
Genomic Map