siRNA Details
| Gene Symbol: MTMR7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004686.4 | NP_004677.3 | 3 | 454 | ||
| XM_017013956.1 | XP_016869445.1 | 3 | 276 | |||
| GenBank mRNA | AK294468.1 | - | 3 | 215 | ||
| AK316022.1 | - | 3 | 264 | |||
| AK316283.1 | - | 3 | 274 | |||
| BC130577.1 | - | 3 | 240 | 40146608 | ||
| BC130579.1 | - | 3 | 240 | 40146610 | ||
| CR749240.1 | - | 3 | 252 | |||

MTMR7
Genomic Map