siRNA Details
| Gene Symbol: MTMR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004687.4 | NP_004678.3 | 16 | 2719 | ||
| XM_005257784.2 | XP_005257841.1 | 15 | 2868 | |||
| XM_005257785.4 | XP_005257842.1 | 17 | 2836 | |||
| XM_005257786.4 | XP_005257843.1 | 16 | 3623 | |||
| XM_006722168.3 | XP_006722231.1 | 16 | 2663 | |||
| XM_011525460.2 | XP_011523762.1 | 17 | 3716 | |||
| GenBank mRNA | AB014547.2 | - | 14 | 2609 | ||
| AF264717.1 | - | 16 | 2662 | |||
| BC035609.1 | - | 16 | 2680 | 5201989 | ||

MTMR4
Genomic Map