siRNA Details
| Gene Symbol: FAM50A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004699.3 | NP_004690.1 | 11, 12 | 1010 | ||
| GenBank mRNA | AD001530.1 | - | 11, 12 | 975 | ||
| AK293119.1 | - | 11, 12 | 964 | |||
| AK313697.1 | - | 11, 12 | 958 | |||
| BC000028.2 | - | 11, 12 | 981 | 3505365 | ||
| BC015499.2 | - | 1, 2 | 71 | |||
| BQ018378.1 | - | 2, 3 | 356 | |||
| BU679052.1 | - | 2, 3 | 356 | |||
| D83260.1 | - | 11, 12 | 880 | |||

FAM50A
Genomic Map