siRNA Details
| Gene Symbol: KCNQ4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004700.3 | NP_004691.2 | 2, 3 | 487 | ||
| NM_172163.2 | NP_751895.1 | 2, 3 | 487 | |||
| GenBank mRNA | AF105202.1 | - | 2, 3 | 487 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCNQ4| Gene Symbol: KCNQ4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004700.3 | NP_004691.2 | 2, 3 | 487 | ||
| NM_172163.2 | NP_751895.1 | 2, 3 | 487 | |||
| GenBank mRNA | AF105202.1 | - | 2, 3 | 487 | ||