siRNA Details
Gene Symbol: CCNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004701.3 | NP_004692.1 | 7 | 1135 | ||
GenBank mRNA | AB020981.1 | - | 7 | 1020 | ||
AF002822.1 | - | 7 | 970 | |||
AF087910.1 | - | 7 | 1077 | |||
AK001404.1 | - | 7 | 1099 | |||
AK222611.1 | - | 7 | 1068 | |||
AL080146.1 | - | 7 | 1088 | |||
AM851054.1 | - | 7 | 951 | |||
BC105086.1 | - | 7 | 1083 | 8144089 | ||
BC105112.1 | - | 7 | 1083 | 8144115 | ||
BT019458.1 | - | 7 | 951 | |||
CR533527.1 | - | 7 | 951 |
