siRNA Details
| Gene Symbol: CTDP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001202504.1 | NP_001189433.1 | 6, 7 | 637 | ||
| NM_001318511.1 | NP_001305440.1 | 6, 7 | 1010 | |||
| NM_004715.4 | NP_004706.3 | 6, 7 | 1010 | |||
| NM_048368.3 | NP_430255.2 | 6, 7 | 1010 | |||
| XM_011526261.1 | XP_011524563.1 | 6, 7 | 988 | |||
| XM_017026078.1 | XP_016881567.1 | 4, 5 | 490 | |||
| GenBank mRNA | AF081287.1 | - | 5, 6 | 558 | ||
| AF154115.1 | - | 6, 7 | 946 | |||
| BC032515.1 | - | 6, 7 | 664 | |||
| BC047999.1 | - | 6, 7 | 1010 | |||
| BC052576.1 | - | 6, 7 | 602 | |||
| BC063447.1 | - | 6, 7 | 965 | 5500032 | ||

CTDP1
Genomic Map