siRNA Details
| Gene Symbol: MYOM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003970.3 | NP_003961.3 | 11, 12 | 1406 | ||
| XM_006716237.1 | XP_006716300.1 | 11, 12 | 1371 | |||
| GenBank mRNA | BC052969.1 | - | 11, 12 | 1359 | 6205359 | |
| X69089.1 | - | 11, 12 | 1316 | |||

MYOM2
Genomic Map