siRNA Details
| Gene Symbol: ZMYM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171162.1 | NP_001164633.1 | 20 | 3892 | ||
| NM_005096.3 | NP_005087.1 | 20 | 3367 | |||
| NM_201599.2 | NP_963893.1 | 20 | 3358 | |||
| XM_005262309.3 | XP_005262366.1 | 20 | 3801 | |||
| XM_005262310.2 | XP_005262367.1 | 20 | 3278 | |||
| XM_011531062.2 | XP_011529364.1 | 20 | 3267 | |||
| GenBank mRNA | AB002383.1 | - | 20 | 3266 | ||
| BC069057.1 | - | 20 | 3271 | 5499547 | ||
| X95808.1 | - | 21 | 3868 | |||

ZMYM3
Genomic Map