siRNA Details
| Gene Symbol: LARGE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004737.4 | NP_004728.1 | 11 | 1795 | ||
| NM_133642.3 | NP_598397.1 | 10 | 1732 | |||
| XM_005261831.3 | XP_005261888.1 | 11 | 1641 | |||
| XM_005261832.3 | XP_005261889.1 | 10 | 1596 | |||
| XM_011530510.2 | XP_011528812.1 | 11 | 1582 | |||
| XM_011530512.2 | XP_011528814.1 | 6 | 719 | |||
| XM_011530513.2 | XP_011528815.1 | 2 | 260 | |||
| GenBank mRNA | AJ007583.1 | - | 11 | 1705 | ||
| AK294766.1 | - | 10 | 1418 | |||
| AK295636.1 | - | 10 | 1258 | |||
| AK301808.1 | - | 6 | 710 | |||
| AK310462.1 | - | 6 | 719 | |||
| BC117425.1 | - | 9 | 1284 | 40125976 | ||
| BC126404.1 | - | 9 | 1284 | 8992120 | ||
| CR456510.1 | - | 9 | 1244 | |||
| KJ534877.1 | - | 9 | 1216 | |||
| KJ535033.1 | - | 9 | 1216 | |||
| KJ535073.1 | - | 8 | 1092 | |||
| KJ535074.1 | - | 7 | 1007 | |||

LARGE1
Genomic Map