siRNA Details
| Gene Symbol: PNMA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006029.4 | NP_006020.4 | 1 | 838 | ||
| GenBank mRNA | AF320308.1 | - | 1 | 323 | ||
| AK290980.1 | - | 1 | 175 | |||
| BC039577.1 | - | 1 | 141 | 4431875 | ||
| CR457112.1 | - | 1 | 52 | |||

PNMA1
Genomic Map