siRNA Details
| Gene Symbol: WDR46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005452.5 | NP_005443.3 | 1, 2 | 426 | ||
| XM_017011484.1 | XP_016866973.1 | 1, 2 | 140 | |||
| GenBank mRNA | AF447870.1 | - | 1, 2 | 118 | ||
| AK292171.1 | - | 1, 2 | 409 | |||
| AK292616.1 | - | 1, 2 | 209 | |||
| AK311388.1 | - | 1, 2 | 137 | |||
| AK315616.1 | - | 1, 2 | 138 | |||
| BC000388.2 | - | 1, 2 | 113 | 2821198 | ||
| BP344599.1 | - | 1, 2 | 426 | |||
| BX439421.2 | - | 1, 2 | 113 | |||
| DB272479.1 | - | 1, 2 | 137 | |||

WDR46
Genomic Map