siRNA Details
| Gene Symbol: TAAR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033080.1 | NP_001028252.1 | 2 | 197 | ||
| NM_014626.3 | NP_055441.2 | 1 | 85 | |||
| GenBank mRNA | AY702304.1 | - | 1 | 62 | ||
| AY703480.1 | - | 2 | 197 | |||
| BC067461.1 | - | 1 | 62 | |||
| BC067462.1 | - | 1 | 62 | |||
| BC067463.1 | - | 1 | 62 | |||
| DN693314.1 | - | 1 | 85 | |||
More Information
Set Membership: |
|

TAAR2
Genomic Map