siRNA Details
| Gene Symbol: ZNHIT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281433.1 | NP_001268362.1 | 5 | 442 | ||
| NM_001281434.1 | NP_001268363.1 | 3 | 308 | |||
| NM_004773.3 | NP_004764.1 | 5 | 476 | |||
| NR_104009.1 | 5 | 471 | ||||
| NR_104010.1 | 4 | 444 | ||||
| GenBank mRNA | AB209193.1 | - | 3 | 3940 | ||
| AF400652.1 | - | 5 | 468 | |||
| AK290858.1 | - | 5 | 427 | |||
| BC017931.1 | - | 5 | 441 | 4669658 | ||
| BC105608.1 | - | 4 | 416 | |||
| BG032116.1 | - | 5 | 377 | |||
| BM457955.1 | - | 406 | ||||
| BQ009986.1 | - | 1 | 495 | |||
| CB996852.1 | - | 5 | 502 | |||
| L40410.1 | - | 5 | 394 | |||

ZNHIT3
Genomic Map