siRNA Details
| Gene Symbol: TGM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004245.3 | NP_004236.1 | 9 | 1373 | ||
| NM_201631.3 | NP_963925.2 | 10 | 1619 | |||
| XM_011522230.2 | XP_011520532.1 | 4 | 801 | |||
| XM_017022729.1 | XP_016878218.1 | 4 | 825 | |||
| GenBank mRNA | AF035960.1 | - | 10 | 1619 | ||
| AF035961.1 | - | 9 | 1373 | |||
| AK298479.1 | - | 6 | 1035 | |||
| BC119009.1 | - | 10 | 1619 | 40085374 | ||
| BC122859.1 | - | 10 | 1619 | 40085372 | ||

TGM5
Genomic Map