siRNA Details
| Gene Symbol: RAB33A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004794.2 | NP_004785.1 | 2 | 669 | ||
| XM_017029963.1 | XP_016885452.1 | 3 | 358 | |||
| GenBank mRNA | AF498959.1 | - | 2 | 405 | ||
| AK094927.1 | - | 3 | 1347 | |||
| BC001157.1 | - | 2 | 669 | 3510745 | ||
| BC009996.2 | - | 2 | 669 | 4129324 | ||
| BT007038.1 | - | 2 | 405 | |||
| D14889.1 | - | 2 | 842 | |||

RAB33A
Genomic Map