siRNA Details
| Gene Symbol: RAB33A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004794.2 | NP_004785.1 | 2 | 770 | ||
| XM_017029963.1 | XP_016885452.1 | 3 | 459 | |||
| GenBank mRNA | AF498959.1 | - | 2 | 506 | ||
| AK094927.1 | - | 3 | 1448 | |||
| BC001157.1 | - | 2 | 770 | 3510745 | ||
| BC009996.2 | - | 2 | 770 | 4129324 | ||
| BT007038.1 | - | 2 | 506 | |||
| D14889.1 | - | 2 | 943 | |||

RAB33A
Genomic Map