siRNA Details
| Gene Symbol: CIAO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004804.2 | NP_004795.1 | 4, 5 | 701 | ||
| GenBank mRNA | AK056423.1 | - | 3, 4 | 458 | ||
| AK223257.1 | - | 4, 5 | 622 | |||
| AK299503.1 | - | 4, 5 | 388 | |||
| BC001395.2 | - | 4, 5 | 565 | 3050587 | ||
| BC032812.1 | - | 4, 5 | 595 | 5219740 | ||
| CR456802.1 | - | 4, 5 | 496 | |||
| EF011618.1 | - | 4, 5 | 595 | |||
| U63810.1 | - | 4, 5 | 652 | |||

CIAO1
Genomic Map