siRNA Details
| Gene Symbol: QKI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301085.1 | NP_001288014.1 | 4 | 1070 | ||
| NM_006775.2 | NP_006766.1 | 4 | 1070 | |||
| NM_206853.2 | NP_996735.1 | 4 | 1070 | |||
| NM_206854.2 | NP_996736.1 | 4 | 1070 | |||
| NM_206855.2 | NP_996737.1 | 4 | 1070 | |||
| XM_011536258.2 | XP_011534560.1 | 4 | 577 | |||
| XM_011536259.2 | XP_011534561.1 | 4 | 577 | |||
| XM_011536261.1 | XP_011534563.1 | 4 | 577 | |||
| XM_017011502.1 | XP_016866991.1 | 4 | 577 | |||
| XM_017011503.1 | XP_016866992.1 | 4 | 577 | |||
| XM_017011504.1 | XP_016866993.1 | 4 | 577 | |||
| GenBank mRNA | AB067798.1 | - | 4 | 1071 | ||
| AB067799.1 | - | 4 | 1071 | |||
| AB067800.1 | - | 4 | 1071 | |||
| AB067801.1 | - | 4 | 1071 | |||
| AF142422.1 | - | 4 | 630 | |||
| AF272349.1 | - | 4 | 952 | |||
| AK027309.1 | - | 4 | 477 | |||
| AK295235.1 | - | 1 | 81 | |||
| AK308784.1 | - | 1 | 18 | |||
| AY780788.1 | - | 4 | 507 | |||
| BC012222.1 | - | 3 | 377 | |||
| BC019917.2 | - | 4 | 558 | 3921310 | ||
| DQ323998.1 | - | 4 | 519 | |||

QKI
Genomic Map