siRNA Details
| Gene Symbol: RASAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004841.3 | NP_004832.1 | 10 | 1877 | ||
| NM_170692.2 | NP_733793.2 | 12 | 2960 | |||
| XM_005245622.3 | XP_005245679.1 | 10 | 2533 | |||
| XM_011510166.2 | XP_011508468.1 | 12 | 2960 | |||
| XM_011510167.2 | XP_011508469.1 | 12 | 2960 | |||
| XM_017002849.1 | XP_016858338.1 | 12 | 5566 | |||
| XM_017002850.1 | XP_016858339.1 | 12 | 5565 | |||
| XM_017002851.1 | XP_016858340.1 | 12 | 5565 | |||
| XM_017002852.1 | XP_016858341.1 | 12 | 5564 | |||
| XM_017002853.1 | XP_016858342.1 | 12 | 3004 | |||
| XM_017002854.1 | XP_016858343.1 | 10 | 2536 | |||
| XM_017002855.1 | XP_016858344.1 | 10 | 2535 | |||
| GenBank mRNA | AF047711.1 | - | 10 | 1877 | ||
| AK022662.1 | - | 2 | 403 | |||
| AK290308.1 | - | 10 | 1820 | |||
| BC110611.2 | - | 12 | 2192 | 40028914 | ||

RASAL2
Genomic Map