siRNA Details
| Gene Symbol: EIF4E2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276336.1 | NP_001263265.1 | 2, 3 | 267 | ||
| NM_001282958.1 | NP_001269887.1 | 2, 3 | 267 | |||
| NM_004846.3 | NP_004837.1 | 2, 3 | 267 | |||
| XM_011512206.2 | XP_011510508.1 | 2, 3 | 542 | |||
| GenBank mRNA | AB209519.1 | - | 2, 3 | 171 | ||
| AF038957.1 | - | 2, 3 | 160 | |||
| AF047695.1 | - | 2, 3 | 158 | |||
| AF068117.1 | - | 2, 3 | 143 | |||
| AK022518.1 | - | 2, 3 | 165 | |||
| BC005392.1 | - | 2, 3 | 165 | 3997986 | ||
| BC005874.2 | - | 2, 3 | 187 | 2961437 | ||
| BC021226.2 | - | 2, 3 | 131 | 4302323 | ||
| BC021690.1 | - | 2, 3 | 169 | 3961350 | ||
| BG913475.1 | - | 2, 3 | 168 | |||
| CB994873.1 | - | 235 | ||||
| DA992544.1 | - | 2, 3 | 267 | |||

EIF4E2
Genomic Map