siRNA Details
| Gene Symbol: STX8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004853.2 | NP_004844.1 | 8 | 823 | ||
| NR_033656.1 | 6 | 617 | ||||
| XM_011524079.2 | XP_011522381.1 | 6 | 890 | |||
| GenBank mRNA | AF036715.1 | - | 8 | 673 | ||
| AF062077.1 | - | 8 | 823 | |||
| AF115323.1 | - | 8 | 823 | |||
| BC009713.1 | - | 8 | 696 | 3899766 | ||
| BT007319.1 | - | 8 | 673 | |||
| BU596252.1 | - | 6 | 495 | |||

STX8
Genomic Map