siRNA Details
| Gene Symbol: SLC4A8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039960.2 | NP_001035049.1 | 18 | 2556 | ||
| NM_001258401.2 | NP_001245330.1 | 18 | 2502 | |||
| XM_011539010.1 | XP_011537312.1 | 18 | 2557 | |||
| XM_011539011.1 | XP_011537313.1 | 18 | 2502 | |||
| XM_011539014.2 | XP_011537316.1 | 8 | 1155 | |||
| XM_017020240.1 | XP_016875729.1 | 18 | 2584 | |||
| XM_017020241.1 | XP_016875730.1 | 18 | 2916 | |||
| XM_017020242.1 | XP_016875731.1 | 18 | 2571 | |||
| GenBank mRNA | AB018282.1 | - | 20 | 2636 | ||
| AF069512.1 | - | 18 | 2393 | |||
| DQ063579.2 | - | 18 | 2545 | |||
| DQ975204.1 | - | 18 | 2372 | |||
| DQ996398.1 | - | 18 | 2606 | |||
| DQ996537.1 | - | 18 | 2372 | |||

SLC4A8
Genomic Map