siRNA Details
| Gene Symbol: APBA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004886.3 | NP_004877.1 | 5 | 996 | ||
| XM_006722950.3 | XP_006723013.1 | 5 | 1018 | |||
| XM_006722951.3 | XP_006723014.1 | 3 | 703 | |||
| GenBank mRNA | AB021638.1 | - | 5 | 986 | ||
| AB209603.1 | - | 1 | 2225 | |||
| BC008338.1 | - | 3 | 843 | |||
| BC086306.1 | - | 4 | 855 | 6371716 | ||

APBA3
Genomic Map