siRNA Details
| Gene Symbol: TESMIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039656.1 | NP_001034745.1 | 2 | 193 | ||
| NM_004923.3 | NP_004914.2 | 2 | 193 | |||
| XM_011545402.1 | XP_011543704.1 | 2 | 237 | |||
| XM_011545403.2 | XP_011543705.1 | 1 | 106 | |||
| XM_017018588.1 | XP_016874077.1 | 2 | 237 | |||
| GenBank mRNA | AK074817.1 | - | 2 | 140 | ||
| AK292358.1 | - | 2 | 133 | |||
| BC027593.1 | - | 2 | 129 | 4838081 | ||
| BC064579.1 | - | 2 | 160 | 5743865 | ||
| BG772890.1 | - | 2 | 211 | |||
| BM790577.1 | - | 2 | 193 | |||

TESMIN
Genomic Map