siRNA Details
| Gene Symbol: TESMIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039656.1 | NP_001034745.1 | 5 | 906 | ||
| NM_004923.3 | NP_004914.2 | 5 | 906 | |||
| XM_011545402.1 | XP_011543704.1 | 5 | 950 | |||
| XM_011545403.2 | XP_011543705.1 | 4 | 819 | |||
| GenBank mRNA | AK292358.1 | - | 5 | 846 | ||
| BC027593.1 | - | 5 | 842 | 4838081 | ||
| BC064579.1 | - | 5 | 873 | 5743865 | ||
| U86074.1 | - | 4 | 545 | |||

TESMIN
Genomic Map