siRNA Details
| Gene Symbol: ISG15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005101.3 | NP_005092.1 | 2 | 365 | ||
| GenBank mRNA | BM712238.1 | - | 2 | 366 | ||
| BU161226.1 | - | 2 | 322 | |||
| M13755.1 | - | 2 | 333 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ISG15| Gene Symbol: ISG15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005101.3 | NP_005092.1 | 2 | 365 | ||
| GenBank mRNA | BM712238.1 | - | 2 | 366 | ||
| BU161226.1 | - | 2 | 322 | |||
| M13755.1 | - | 2 | 333 | |||