siRNA Details
| Gene Symbol: MDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014641.2 | NP_055456.2 | 14, 15 | 6540 | ||
| XM_005249492.2 | XP_005249549.1 | 14, 15 | 6444 | |||
| XM_005249493.3 | XP_005249550.1 | 14, 15 | 6441 | |||
| XM_005249494.4 | XP_005249551.1 | 14, 15 | 6843 | |||
| XM_005249497.3 | XP_005249554.1 | 14, 15 | 5874 | |||
| XM_005249498.3 | XP_005249555.1 | 13, 14 | 5652 | |||
| XM_011515001.2 | XP_011513303.1 | 14, 15 | 6271 | |||
| XM_011515003.2 | XP_011513305.1 | 14, 15 | 6126 | |||
| XM_011515004.2 | XP_011513306.1 | 15, 16 | 6155 | |||
| XM_017011519.1 | XP_016867008.1 | 14, 15 | 6223 | |||
| XM_017011520.1 | XP_016867009.1 | 14, 15 | 6139 | |||
| XM_017011521.1 | XP_016867010.1 | 14, 15 | 5572 | |||
| XM_017011522.1 | XP_016867011.1 | 13, 14 | 5649 | |||
| GenBank mRNA | BC110645.1 | - | 9, 10 | 2197 | ||
| BC131491.1 | - | 10, 11 | 3511 | |||
| BC152556.1 | - | 14, 15 | 5252 | 9007258 | ||
| CR749828.1 | - | 10, 11 | 4172 | |||
| D79992.1 | - | 13, 14 | 6113 | |||

MDC1
Genomic Map