siRNA Details
| Gene Symbol: CEP135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025009.4 | NP_079285.2 | 10 | 1449 | ||
| XM_005265788.3 | XP_005265845.1 | 10 | 1416 | |||
| XM_006714055.2 | XP_006714118.1 | 10 | 1416 | |||
| GenBank mRNA | AB014535.1 | - | 3 | 1102 | ||
| BC012003.1 | - | 5 | 532 | |||
| BC136535.1 | - | 10 | 1276 | 9020525 | ||
| BC136536.1 | - | 10 | 1276 | 9020526 | ||

CEP135
Genomic Map