siRNA Details
| Gene Symbol: FAM53B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014661.3 | NP_055476.3 | 4 | 1216 | ||
| GenBank mRNA | AK127343.1 | - | 4 | 1410 | ||
| BC031654.1 | - | 4 | 1124 | 5174078 | ||
| D50930.1 | - | 4 | 905 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM53B| Gene Symbol: FAM53B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014661.3 | NP_055476.3 | 4 | 1216 | ||
| GenBank mRNA | AK127343.1 | - | 4 | 1410 | ||
| BC031654.1 | - | 4 | 1124 | 5174078 | ||
| D50930.1 | - | 4 | 905 | |||