siRNA Details
| Gene Symbol: UBE3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014671.2 | NP_055486.2 | 12 | 1833 | ||
| XM_005249564.4 | XP_005249621.1 | 11 | 1767 | |||
| XM_017012818.1 | XP_016868307.1 | 13 | 2105 | |||
| GenBank mRNA | AK295138.1 | - | 12 | 1513 | ||
| BC014029.2 | - | 13 | 1593 | 4541634 | ||
| D13635.1 | - | 12 | 1824 | |||
| DB085748.1 | - | 3 | 363 | |||

UBE3C
Genomic Map