siRNA Details
| Gene Symbol: TRAM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012288.3 | NP_036420.1 | 4 | 508 | ||
| XM_011515005.2 | XP_011513307.1 | 4 | 383 | |||
| GenBank mRNA | AB621814.1 | - | 4 | 504 | ||
| AK291751.1 | - | 4 | 394 | |||
| AK311560.1 | - | 4 | 377 | |||
| BC028121.1 | - | 4 | 508 | 5169808 | ||
| D31762.1 | - | 4 | 434 | |||
| KU178488.1 | - | 4 | 359 | |||
| KU178489.1 | - | 3 | 295 | |||

TRAM2
Genomic Map