siRNA Details
| Gene Symbol: TRAM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NM_012288.3 | NP_036420.1 | 9 | 1000 | ||
| XM_011515005.2 | XP_011513307.1 | 9 | 875 | |||
| GenBank mRNA | AK291751.1 | - | 9 | 886 | ||
| AK311560.1 | - | 9 | 869 | |||
| AY927606.1 | - | LIT3596 AY927606 |
2 | 169 | ||
| BC028121.1 | - | 9 | 1000 | 5169808 | ||
| D31762.1 | - | 9 | 926 | |||
| KU178488.1 | - | 9 | 851 | |||
| KU178489.1 | - | 8 | 787 | |||

TRAM2
Genomic Map