siRNA Details
| Gene Symbol: CEP104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014704.3 | NP_055519.1 | 10 | 1516 | ||
| XM_005244815.4 | XP_005244872.1 | 11 | 1616 | |||
| XM_011542473.2 | XP_011540775.1 | 11 | 1816 | |||
| XM_011542474.2 | XP_011540776.1 | 10 | 1534 | |||
| XM_011542475.2 | XP_011540777.1 | 11 | 1642 | |||
| XM_011542478.1 | XP_011540780.1 | 11 | 1816 | |||
| XM_017002917.1 | XP_016858406.1 | 11 | 1624 | |||
| XM_017002918.1 | XP_016858407.1 | 10 | 1342 | |||
| GenBank mRNA | AB011134.1 | - | 10 | 1470 | ||
| AK308191.1 | - | 10 | 1474 | |||
| BC001640.2 | - | 10 | 1481 | |||

CEP104
Genomic Map