siRNA Details
| Gene Symbol: LAPTM4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014713.4 | NP_055528.1 | 3, 4 | 816 | ||
| GenBank mRNA | AK129875.1 | - | 3, 4 | 466 | ||
| AK304433.1 | - | 3, 4 | 384 | |||
| AY359028.1 | - | 3, 4 | 432 | |||
| BC000421.2 | - | 3, 4 | 393 | 2964450 | ||
| BC093075.1 | - | 3, 4 | 416 | 30402963 | ||
| BP354016.1 | - | 3, 4 | 541 | |||
| CB962971.1 | - | 478 | ||||
| CR456758.1 | - | 3, 4 | 308 | |||
| D14696.1 | - | 3, 4 | 454 | |||

LAPTM4A
Genomic Map