siRNA Details
| Gene Symbol: SLK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304743.1 | NP_001291672.1 | 9 | 1883 | ||
| NM_014720.3 | NP_055535.2 | 9 | 1883 | |||
| GenBank mRNA | AB002804.1 | - | 9 | 1554 | ||
| AF273048.1 | - | 9 | 1356 | |||
| AK302855.1 | - | 9 | 1316 | |||
| AK307389.1 | - | 10 | 1435 | |||
| BC047762.1 | - | 9 | 1876 | |||
| BC064804.1 | - | 9 | 1879 | |||
| BC111565.1 | - | 9 | 1353 | 40009070 | ||
| D86959.1 | - | 9 | 1833 | |||

SLK
Genomic Map