siRNA Details
| Gene Symbol: FRMPD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014728.3 | NP_055543.2 | 4 | 861 | ||
| XM_005274632.3 | XP_005274689.2 | 4 | 539 | |||
| XM_011545613.2 | XP_011543915.1 | 4 | 848 | |||
| XM_017029983.1 | XP_016885472.1 | 4 | 751 | |||
| XM_017029984.1 | XP_016885473.1 | 4 | 540 | |||
| XM_017029985.1 | XP_016885474.1 | 4 | 537 | |||
| XM_017029986.1 | XP_016885475.1 | 4 | 750 | |||
| GenBank mRNA | AB002314.3 | - | 4 | 861 | ||
| AK289694.1 | - | 4 | 848 | |||
| BC113700.1 | - | 4 | 446 | 8322752 | ||
| BC113702.1 | - | 4 | 446 | 8322754 | ||
| BC143746.1 | - | 5 | 543 | 9052260 | ||

FRMPD4
Genomic Map