siRNA Details
| Gene Symbol: FRMPD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014728.3 | NP_055543.2 | 5, 6 | 981 | ||
| XM_005274632.3 | XP_005274689.2 | 5, 6 | 659 | |||
| XM_011545613.2 | XP_011543915.1 | 5, 6 | 968 | |||
| XM_017029983.1 | XP_016885472.1 | 5, 6 | 871 | |||
| XM_017029984.1 | XP_016885473.1 | 5, 6 | 660 | |||
| XM_017029985.1 | XP_016885474.1 | 5, 6 | 657 | |||
| XM_017029986.1 | XP_016885475.1 | 5, 6 | 870 | |||
| GenBank mRNA | AB002314.3 | - | 5, 6 | 981 | ||
| AK289694.1 | - | 5, 6 | 968 | |||
| BC113700.1 | - | 5, 6 | 566 | 8322752 | ||
| BC113702.1 | - | 5, 6 | 566 | 8322754 | ||
| BC143746.1 | - | 6, 7 | 663 | 9052260 | ||

FRMPD4
Genomic Map