siRNA Details
| Gene Symbol: SUSD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014734.3 | NP_055549.1 | 3, 4 | 628 | ||
| XM_011537412.1 | XP_011535714.1 | 3, 4 | 529 | |||
| XM_011537413.1 | XP_011535715.1 | 4, 5 | 815 | |||
| XM_017021838.1 | XP_016877327.1 | 3, 4 | 518 | |||
| XM_017021839.1 | XP_016877328.1 | 3, 4 | 414 | |||
| GenBank mRNA | AK309302.1 | - | 4, 5 | 821 | ||
| BC064697.1 | - | 3, 4 | 625 | 30344263 | ||
| D87434.1 | - | 3, 4 | 580 | |||

SUSD6
Genomic Map