siRNA Details
Gene Symbol: RNF144A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014746.4 | NP_055561.2 | 9 | 1291 | ||
XM_005246200.3 | XP_005246257.1 | 9 | 2482 | |||
XM_005246202.4 | XP_005246259.1 | 8 | 1122 | |||
XM_017005396.1 | XP_016860885.1 | 9 | 2458 | |||
XM_017005397.1 | XP_016860886.1 | 9 | 1116 | |||
XM_017005404.1 | XP_016860893.1 | 8 | 1099 | |||
GenBank mRNA | BC050373.1 | - | 9 | 1231 | 5722878 | |
D79983.1 | - | 9 | 1197 |
