siRNA Details
| Gene Symbol: KIAA0586 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001244189.1 | NP_001231118.1 | 29, 30 | 4569 | ||
| NM_001244190.1 | NP_001231119.1 | 28, 29 | 4412 | |||
| NM_001244191.1 | NP_001231120.1 | 27, 28 | 4220 | |||
| NM_001244192.1 | NP_001231121.1 | 28, 29 | 4343 | |||
| NM_001244193.1 | NP_001231122.1 | 24, 25 | 3856 | |||
| NM_014749.3 | NP_055564.3 | 26, 27 | 4211 | |||
| GenBank mRNA | AB011158.1 | - | 28, 29 | 4395 | ||
| AK302579.1 | - | 24, 25 | 3858 | |||
| AK302635.1 | - | 27, 28 | 4179 | |||
| AK302836.1 | - | 29, 30 | 4569 | |||
| AY359881.1 | - | 28, 29 | 4292 | |||
| BC000900.2 | - | 4, 5 | 564 | |||
| BC066647.1 | - | 26, 27 | 4166 | 6166031 | ||

KIAA0586
Genomic Map