siRNA Details
| Gene Symbol: SCRN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145513.1 | NP_001138985.1 | 3 | 374 | ||
| NM_001145514.1 | NP_001138986.1 | 3 | 306 | |||
| NM_001145515.1 | NP_001138987.1 | 2 | 206 | |||
| NM_014766.4 | NP_055581.3 | 3 | 366 | |||
| GenBank mRNA | AB071705.1 | - | 3 | 256 | ||
| AK289514.1 | - | 2 | 401 | |||
| AK293462.1 | - | 3 | 318 | |||
| AK295172.1 | - | 3 | 306 | |||
| AK295713.1 | - | 2 | 109 | |||
| BC040492.1 | - | 3 | 318 | 5314711 | ||
| D83777.2 | - | 3 | 311 | |||
| DC307846.1 | - | 3 | 375 | |||
| DC370097.1 | - | 3 | 366 | |||

SCRN1
Genomic Map