siRNA Details
| Gene Symbol: KIAA0141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142603.2 | NP_001136075.1 | 3 | 321 | ||
| NM_014773.4 | NP_055588.3 | 3 | 321 | |||
| XM_005268547.3 | XP_005268604.1 | 3 | 356 | |||
| XM_005268548.3 | XP_005268605.1 | 3 | 356 | |||
| XM_005268552.2 | XP_005268609.1 | 3 | 356 | |||
| XM_011537715.1 | XP_011536017.1 | 3 | 356 | |||
| XM_011537716.2 | XP_011536018.1 | 3 | 356 | |||
| GenBank mRNA | AK292869.1 | - | 3 | 270 | ||
| BC007855.2 | - | 3 | 297 | 4298364 | ||
| BC010452.1 | - | 3 | 212 | 4155782 | ||
| BC011269.1 | - | 3 | 212 | 4155755 | ||
| BC011926.1 | - | 3 | 242 | 4543650 | ||
| BC035486.1 | - | 3 | 237 | |||
| CN307416.1 | - | 3 | 357 | |||
| D50931.1 | - | 3 | 279 | |||

KIAA0141
Genomic Map