siRNA Details
| Gene Symbol: EFCAB14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014774.2 | NP_055589.1 | 7 | 1866 | ||
| GenBank mRNA | AB007963.1 | - | 7 | 1866 | ||
| AK001487.1 | - | 4 | 365 | |||
| BC002525.2 | - | 7 | 1011 | 3139273 | ||
| BC018842.2 | - | 7 | 1011 | |||

EFCAB14
Genomic Map