siRNA Details
| Gene Symbol: CUL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168370.1 | NP_001161842.1 | 4 | 1101 | ||
| NM_014780.4 | NP_055595.2 | 4 | 1094 | |||
| XM_005249503.2 | XP_005249560.1 | 4 | 957 | |||
| XM_006715285.1 | XP_006715348.1 | 4 | 1195 | |||
| XM_011515019.2 | XP_011513321.1 | 4 | 1052 | |||
| XM_011515020.2 | XP_011513322.1 | 4 | 955 | |||
| XM_017011533.1 | XP_016867022.1 | 4 | 1053 | |||
| XM_017011534.1 | XP_016867023.1 | 4 | 1052 | |||
| XM_017011535.1 | XP_016867024.1 | 4 | 956 | |||
| XM_017011536.1 | XP_016867025.1 | 4 | 1195 | |||
| XM_017011537.1 | XP_016867026.1 | 4 | 1195 | |||
| XM_017011538.1 | XP_016867027.1 | 4 | 1099 | |||
| XM_017011539.1 | XP_016867028.1 | 4 | 1099 | |||
| XM_017011540.1 | XP_016867029.1 | 4 | 1058 | |||
| GenBank mRNA | AK292806.1 | - | 4 | 1031 | ||
| AK302668.1 | - | 4 | 1038 | |||
| BC033647.1 | - | 4 | 1076 | 5580027 | ||
| D38548.1 | - | 4 | 848 | |||

CUL7
Genomic Map