siRNA Details
| Gene Symbol: SPATA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135773.1 | NP_001129245.1 | 3 | 872 | ||
| NM_006038.3 | NP_006029.1 | 3 | 788 | |||
| XM_006723894.1 | XP_006723957.1 | 3 | 708 | |||
| XM_011529116.1 | XP_011527418.1 | 3 | 745 | |||
| GenBank mRNA | AB018300.1 | - | 3 | 706 | ||
| AK023229.1 | - | 3 | 743 | |||
| AK289644.1 | - | 3 | 872 | |||
| AK295536.1 | - | 2 | 350 | |||
| BC009481.2 | - | 3 | 695 | 3953522 | ||

SPATA2
Genomic Map