siRNA Details
| Gene Symbol: ZBTB24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014797.2 | NP_055612.2 | 7 | 1739 | ||
| GenBank mRNA | AB007901.1 | - | 7 | 1739 | ||
| AK309697.1 | - | 7 | 1754 | |||
| BC117374.1 | - | 6 | 1607 | 40125925 | ||
| BC117376.1 | - | 6 | 1607 | 40125927 | ||
| BX648883.1 | - | 915 | ||||

ZBTB24
Genomic Map