siRNA Details
| Gene Symbol: ZBTB24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164313.1 | NP_001157785.1 | 2 | 948 | ||
| NM_014797.2 | NP_055612.2 | 2 | 948 | |||
| GenBank mRNA | AB007901.1 | - | 2 | 948 | ||
| AK309697.1 | - | 2 | 902 | |||
| BC036731.1 | - | 2 | 927 | |||
| BC117374.1 | - | 1 | 816 | 40125925 | ||
| BC117376.1 | - | 1 | 816 | 40125927 | ||
| BX648883.1 | - | 292 | ||||

ZBTB24
Genomic Map