siRNA Details
| Gene Symbol: GAB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012296.3 | NP_036428.1 | 4 | 775 | ||
| NM_080491.2 | NP_536739.1 | 4 | 831 | |||
| XM_006718753.2 | XP_006718816.1 | 5 | 876 | |||
| XM_011545408.2 | XP_011543710.1 | 3 | 482 | |||
| GenBank mRNA | AB011143.1 | - | 4 | 775 | ||
| AB018413.1 | - | 4 | 729 | |||
| BC131711.1 | - | 4 | 733 | 40122341 | ||
| BC152459.1 | - | 4 | 775 | 8862595 | ||

GAB2
Genomic Map